missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenol, 97%

Product Code. 11311317
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311317 5 g 5g
11321317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311317 Supplier Thermo Scientific Alfa Aesar Supplier No. L01527.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 642-71-7 | C9H12O4 | 184.191 g/mol

3,4,5-Trimethoxyphenol is used as Flavonoid drug intermediates. It is used for pharmaceutical, cosmetics, agrochemicals and food industry research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxyphenol is used as Flavonoid drug intermediates. It is used for pharmaceutical, cosmetics, agrochemicals and food industry research.

Solubility
Soluble in chloroform.

Notes
Light Sensitive. Store in the dark. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 642-71-7
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.191
MDL Number MFCD00008389
InChI Key VTCDZPUMZAZMSB-UHFFFAOYSA-N
Synonym antiarol, phenol, 3,4,5-trimethoxy, 3,4,5-trimethoxy phenol, spectrum_000525, specplus_000623, spectrum3_001208, spectrum4_001487, spectrum5_000193, 3,4,5-trirnethoxyphenol, 3,4,5-trimethoxy-phenol
PubChem CID 69505
ChEBI CHEBI:2760
IUPAC Name 3,4,5-trimethoxyphenol
SMILES COC1=CC(=CC(=C1OC)OC)O

Specifications

Melting Point 143°C to 148°C
Quantity 5 g
Beilstein 1958573
Sensitivity Light sensitive
Solubility Information Soluble in chloroform.
Formula Weight 184.19
Percent Purity 97%
Chemical Name or Material 3,4,5-Trimethoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.