missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenol, 98.5+%

Product Code. 11974751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11974751 100 g Glass bottle 100g
10285520 5 g Glass bottle 5g
10295520 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11974751 Supplier Thermo Scientific Acros Supplier No. 189141000
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 642-71-7 | C9H12O4 | 184.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 642-71-7
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.19
MDL Number MFCD00008389
InChI Key VTCDZPUMZAZMSB-UHFFFAOYSA-N
Synonym antiarol, phenol, 3,4,5-trimethoxy, 3,4,5-trimethoxy phenol, spectrum_000525, specplus_000623, spectrum3_001208, spectrum4_001487, spectrum5_000193, 3,4,5-trirnethoxyphenol, 3,4,5-trimethoxy-phenol
PubChem CID 69505
ChEBI CHEBI:2760
IUPAC Name 3,4,5-trimethoxyphenol
SMILES COC1=CC(=CC(=C1OC)OC)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 146.0°C to 149.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)3C6H2OH
Quantity 100 g
Beilstein 06,1154
Formula Weight 184.19
Percent Purity 98.5+%
Physical Form Chunks and/or Powder
Chemical Name or Material 3, 4, 5-Trimethoxyphenol, 98.5+%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4,5-Trimethoxyphenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.