missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenylacetic acid, 99%

Product Code. 11414627
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11414627 5 g 5g
11424627 25 g 25g
11434627 100 g 100g
3 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 11414627

missing translation for 'mfr': Thermo Scientific Alfa Aesar B21136.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 951-82-6 | C11H14O5 | 226.228 g/mol

It is used in the synthesis of 3,4-Diaryl-2(5H)-furanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of 3,4-Diaryl-2(5H)-furanones.

Solubility
Soluble in water. Solubility in methanol is almost transparent.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 951-82-6
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.228
MDL Number MFCD00004336
InChI Key DDSJXCGGOXKGSJ-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyphenylacetic acid, 2-3,4,5-trimethoxyphenyl acetic acid, 3,4,5-trimethoxyphenyl acetic acid, benzeneacetic acid, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylaceticacid, pubchem14015, acmc-209rv8, ksc487g4j, 3,4,5-trimethoxybenzeneacetic acid, benzeneacetic acid,4,5-trimethoxy
PubChem CID 70372
IUPAC Name 2-(3,4,5-trimethoxyphenyl)acetic acid
SMILES COC1=CC(=CC(=C1OC)OC)CC(=O)O

Specifications

Melting Point 118°C to 122°C
Quantity 5 g
Beilstein 2697844
Solubility Information Soluble in water. Solubility in methanol is almost transparent.
Formula Weight 226.23
Percent Purity 99%
Chemical Name or Material 3,4,5-Trimethoxyphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.