missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenylacetic acid, 99%

Product Code. 10761915 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10761915 5 g Glass bottle 5g
10592531 10 g Glass bottle 10g
10676524 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10761915 Supplier Thermo Scientific Acros Supplier No. 139990050
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 951-82-6 | C11H14O5 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 951-82-6
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.23
MDL Number MFCD00004336
InChI Key DDSJXCGGOXKGSJ-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyphenylacetic acid, 2-3,4,5-trimethoxyphenyl acetic acid, 3,4,5-trimethoxyphenyl acetic acid, benzeneacetic acid, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylaceticacid, pubchem14015, acmc-209rv8, ksc487g4j, 3,4,5-trimethoxybenzeneacetic acid, benzeneacetic acid,4,5-trimethoxy
PubChem CID 70372
IUPAC Name 2-(3,4,5-trimethoxyphenyl)acetic acid
SMILES COC1=CC(=CC(=C1OC)OC)CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 117.0°C to 120.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3O)3C6H2CH2CO2H
Quantity 5 g
Beilstein 10, 492
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone and in chloroform
Formula Weight 226.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4, 5-Trimethoxyphenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,4,5-Trimethoxyphenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.