missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenylglyoxal hydrate, Thermo Scientific™

Product Code. 10537580
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10537580 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10537580 Supplier Thermo Scientific Maybridge Supplier No. MAY00159DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 150114-69-5
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Molecular Formula C11H12O5
MDL Number MFCD13151985
Quantity 1 g
Synonym 3,4,5-trimethoxyphenylglyoxal hydrate, 2-oxo-2-3,4,5-trimethoxyphenyl acetaldehyde hydrate, acmc-20ang0, bestipharma 543-750, benzeneacetaldehyde,3,4,5-trimethoxy-a-oxo, oxo 3,4,5-trimethoxyphenyl acetaldehyde-water 1/1, 2-oxidanylidene-2-3,4,5-trimethoxyphenyl ethanal hydrate
InChI Key OWONHIWZFXFPFS-UHFFFAOYSA-N
SMILES COC1=CC(=CC(OC)=C1OC)C(=O)C=O
IUPAC Name 2-oxo-2-(3,4,5-trimethoxyphenyl)acetaldehyde;hydrate
Molecular Weight (g/mol) 224.21
PubChem CID 45074958
Formula Weight 242.23
Percent Purity 97%
Chemical Name or Material 3, 4, 5-Trimethoxyphenylglyoxal hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.