missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethylphenol, 97%

Product Code. 11357895
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357895 25 g 25g
11337895 1 g 1g
11347895 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357895 Supplier Thermo Scientific Alfa Aesar Supplier No. L02911.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 527-54-8 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-54-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002305
InChI Key FDQQNNZKEJIHMS-UHFFFAOYSA-N
PubChem CID 10696
ChEBI CHEBI:38896
IUPAC Name 3,4,5-trimethylphenol
SMILES CC1=CC(=CC(=C1C)C)O

Specifications

Melting Point 104°C to 108°C
Odor Phenol-like
Quantity 25 g
UN Number UN2430
Beilstein 1907246
Synonym phenol, 3,4,5-trimethyl, 3,4,5-hemimellitenol, 1-hydroxy-3,4,5-trimethylbenzene, 5-hydroxy-1,2,3-trimethylbenzene, phenol,3,4,5-trimethyl, 3,5-hemimellitenol, 3,5-trimethylphenol, 3,4,5-trimethyl phenol, 3,4,5-trimethyl-phenol, acmc-1bhm2
Formula Weight 136.2
Percent Purity 97%
Chemical Name or Material 3,4,5-Trimethylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.