missing translation for 'onlineSavingsMsg'
Learn More

3',4',7,8-Tetramethoxyflavone, 97%

Product Code. 11313277
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313277 250 mg 250mg
11303277 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313277 Supplier Thermo Scientific Alfa Aesar Supplier No. L13829.MD
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 65548-55-2 | C19H18O6 | 342.347 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65548-55-2
Molecular Formula C19H18O6
Molecular Weight (g/mol) 342.347
MDL Number MFCD00143001
InChI Key ZRLWYUNKZNRQLO-UHFFFAOYSA-N
Synonym 3',4',7,8-tetramethoxyflavone, 2-3,4-dimethoxyphenyl-7,8-dimethoxy-4h-chromen-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxychromen-4-one, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxy, 2-3,4-dimethoxy-phenyl-7,8-dimethoxy-chromen-4-one, acmc-1b8pz, 7,8,3',4'-tetramethoxyflavone, 7,8-dimethoxy-2-3,4-dimethoxyphenyl-4h-chromen-4-one
PubChem CID 4033898
IUPAC Name 2-(3,4-dimethoxyphenyl)-7,8-dimethoxychromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C=C3)OC)OC)OC

Specifications

Melting Point 206°C to 208°C
Quantity 250 mg
Beilstein 330821
Formula Weight 342.35
Percent Purity 97%
Chemical Name or Material 3',4',7,8-Tetramethoxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.