missing translation for 'onlineSavingsMsg'
Learn More

3,4,7,8-Tetramethyl-1,10-phenanthroline, 98+%

Product Code. 10317690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10317690 1 g Glass bottle 1g
11986611 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10317690 Supplier Thermo Scientific Acros Supplier No. 172220010
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 1660-93-1 | C16H16N2 | 236.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660-93-1
Molecular Formula C16H16N2
Molecular Weight (g/mol) 236.32
InChI Key NPAXPTHCUCUHPT-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 3,4,7,8-tetramethyl, 3,4,7,8-tetramethylphenanthroline, 3,4,7,8-tetramethyl-1, 10-phenanthroline, chembl99384, 3,4,7,8-tetramethyl-1,10 phenanthroline, 3,4,7,8-tetramethylpyridino 3,2-h quinoline, acmc-209dtn, 3,4,7,8-me4phenan, bidd:gt0406, 3,4,7,8-tetramethylphenantroline
PubChem CID 74265
IUPAC Name 3,4,7,8-tetramethyl-1,10-phenanthroline
SMILES CC1=CN=C2C(=C1C)C=CC3=C(C(=CN=C32)C)C

Specifications

Melting Point 277.0°C to 280.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in 1,4-dioxane,acetone,dichloromethane,,n,n-dimethylformamide,n,n-dimethylacetamide,meth,anol
Formula Weight 236.32
Percent Purity 98+%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3,4,7,8-Tetramethyl-1,10-phenanthroline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.