missing translation for 'onlineSavingsMsg'
Learn More

3,4,9,10-Perylenetetracarboxylic dianhydride, 98%

Product Code. 11343506
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343506 50 g 50g
11333506 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343506 Supplier Thermo Scientific Alfa Aesar Supplier No. L14370.18
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 128-69-8 | C24H8O6 | 392.32 g/mol

3,4,9,10-Perylenetetracarboxylic dianhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is mainly used in air dried and baking enamels, epoxy, polyurethane and polyester based paints. It is also used in printing inks for bank and security documents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,9,10-Perylenetetracarboxylic dianhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is mainly used in air dried and baking enamels, epoxy, polyurethane and polyester based paints. It is also used in printing inks for bank and security documents.

Solubility
Soluble in water (0.87g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-69-8
Molecular Formula C24H8O6
Molecular Weight (g/mol) 392.32
MDL Number MFCD00006916
InChI Key CLYVDMAATCIVBF-UHFFFAOYSA-N
Synonym 3,4,9,10-perylenetetracarboxylic dianhydride, pigment red 224, ptcda, perylene-3,4,9,10-tetracarboxylic dianhydride, perylenetetracarboxylic anhydride, anthra 2,1,9-def:6,5,10-d'e'f' diisochromene-1,3,8,10-tetraone, perylo 3,4-cd:9,10-c'd' dipyran-1,3,8,10-tetrone, perylenetetracarboxylic acid dianhydride, 3,4:9,10-perylenetetracarboxylic anhydride
PubChem CID 67191
IUPAC Name 7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
SMILES O=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56

Specifications

Melting Point >300°C
Odor Odorless
Quantity 50 g
Beilstein 57831
Sensitivity Moisture sensitive
Solubility Information Soluble in water (0.87g/L at 20°C).
Formula Weight 392.32
Percent Purity 98%
Chemical Name or Material 3,4,9,10-Perylenetetracarboxylic dianhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.