missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Aminophenyl)propionic acid, 97%, Thermo Scientific™

Product Code. 11334256
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11334256 5 g 5g
11324256 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11334256

Brand: Thermo Scientific Alfa Aesar L19330.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2393-17-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00017118
InChI Key WXOHKMNWMKZMND-UHFFFAOYSA-N
Synonym 3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid
PubChem CID 75451
IUPAC Name 3-(4-aminophenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)N

Specifications

Melting Point 133°C to 135°C
Quantity 5 g
Beilstein 2209840
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material 3-(4-Aminophenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.