missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenoxy)-6-methylpyridazine, 97%, Thermo Scientific™

Product Code. 10384132
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10384132 1 g Amber Glass Bottle 1g
10202352 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10384132 Supplier Thermo Scientific Maybridge Supplier No. CC21310DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 368869-96-9
Molecular Formula C11H9BrN2O
Molecular Weight (g/mol) 265.11
MDL Number MFCD03086135
InChI Key LZEJOINDIKGWQJ-UHFFFAOYSA-N
Synonym 3-4-bromophenoxy-6-methylpyridazine, pyridazine,3-4-bromophenoxy-6-methyl, 3-4-bromanylphenoxy-6-methyl-pyridazine, 4-bromo-1-6-methylpyridazin-3-yloxy benzene
PubChem CID 2776522
IUPAC Name 3-(4-bromophenoxy)-6-methylpyridazine
SMILES CC1=NN=C(OC2=CC=C(Br)C=C2)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 265.11
Percent Purity 95%
Chemical Name or Material 3-(4-Bromophenoxy)-6-methylpyridazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.