missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenyl)-5-methyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Product Code. 10510143
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10510143 1 g 1g
10639705 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10510143 Supplier Thermo Scientific Maybridge Supplier No. CC34810DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 118183-92-9
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
MDL Number MFCD00487341
InChI Key YWBIOYGLRGIJRT-UHFFFAOYSA-N
PubChem CID 823560
IUPAC Name 3-(4-bromophenyl)-5-methyl-1,2,4-oxadiazole
SMILES CC1=NC(=NO1)C2=CC=C(C=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 3-4-bromophenyl-5-methyl-1,2,4-oxadiazole, 4-5-methyl-1,2,4-oxadiazol-3-yl bromobenzene, acmc-1ce2y, 3-4-bromophenyl-5-methyl-1,2,4oxadiazole, 1,2,4-oxadiazole, 3-4-bromophenyl-5-methyl, 1,2,4-oxadiazole,3-4-bromophenyl-5-methyl, 3-4-bromo-phenyl-5-methyl-1,2,4 oxadiazole, 3-4-bromophenyl-5-methyl-1,2,4 oxadiazole, 1-bromo-4-5-methyl-1,2,4-oxadiazol-3-yl benzene
Formula Weight 239.07
Percent Purity 97%
Chemical Name or Material 3-(4-Bromophenyl)-5-methyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.