missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chloro-3-fluorophenyl)propionic acid, 96%

Product Code. 15457137
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457137 5 g 5g
15447137 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457137 Supplier Thermo Scientific Alfa Aesar Supplier No. H34205.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 881189-65-7 | C9H8ClFO2 | 202.609 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 881189-65-7
Molecular Formula C9H8ClFO2
Molecular Weight (g/mol) 202.609
MDL Number MFCD04116058
InChI Key GDXMVKNNJGWZFN-UHFFFAOYSA-N
Synonym 3-4-chloro-3-fluorophenyl propionic acid, 3-4-chloro-3-fluorophenyl propanoic acid, benzenepropanoic acid, 4-chloro-3-fluoro, 3-4-chloro-3-fluoro-phenyl-propionic acid, 4-chloro-3-fluorobenzenepropanoic acid, 3-4-chloro-3-fluorophenyl propionicacid
PubChem CID 4680108
IUPAC Name 3-(4-chloro-3-fluorophenyl)propanoic acid
SMILES C1=CC(=C(C=C1CCC(=O)O)F)Cl

Specifications

Melting Point 80°C to 84°C
Quantity 5 g
Formula Weight 202.61
Percent Purity 96%
Chemical Name or Material 3-(4-Chloro-3-fluorophenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.