missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chloro-3-methylphenyl)propionic acid, 96%

Product Code. 15408305
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408305 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15408305 Supplier Thermo Scientific Alfa Aesar Supplier No. H33072.ME
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1086386-05-1 | C10H11ClO2 | 198.646 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1086386-05-1
Melting Point 88°C to 92°C
Molecular Formula C10H11ClO2
MDL Number MFCD11655590
Quantity 500 mg
Synonym 3-4-chloro-3-methylphenyl propanoic acid, 3-4-chloro-3-methyl-phenyl-propionic acid, 3-4-chloro-3-methylphenyl propionic acid, benzenepropanoic acid, 4-chloro-3-methyl
InChI Key PCLIPGMIRVJYFB-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)CCC(=O)O)Cl
IUPAC Name 3-(4-chloro-3-methylphenyl)propanoic acid
Molecular Weight (g/mol) 198.646
PubChem CID 46741400
Formula Weight 198.65
Percent Purity 96%
Chemical Name or Material 3-(4-Chloro-3-methylphenyl)propionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.