missing translation for 'onlineSavingsMsg'
Learn More

3-(4-chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10793053
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10793053 250 mg 250mg
10292972 1 g 1g
10793367 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10793053 Supplier Thermo Scientific Maybridge Supplier No. KM01185CB
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 116525-66-7
Molecular Formula C13H8ClNO2S2
Molecular Weight (g/mol) 309.78
MDL Number MFCD00112478
InChI Key CCQQTQYIJRVXIU-UHFFFAOYSA-N
Synonym 3-4-chlorophenyl-4-cyano-5-methylthio thiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 3-4-chlorophenyl-4-cyano-5-methylthio, 3-4-chlorophenyl-4-cyano-5-methylsulfanyl thiophene-2-carboxylic acid, maybridge1_004446, acmc-1bss2, 3-4-chlorophenyl-4-cyano-5-methylthio-2-thiophenecarboxylic acid, 3-4-chlorophenyl-4-cyano-5-methylsulfanyl-thiophene-2-carboxylic acid, 3-4-chlorophenyl-4-cyano-5-methylsulfanylthiophene-2-carboxylic acid
PubChem CID 2735791
SMILES CSC1=C(C#N)C(=C(S1)C(O)=O)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name 3-(4-chlorophenyl)-4-cyano-5-(methylsulfanyl)thiophene-2-carboxylic acid
Formula Weight 309.8
Percent Purity 97%
Chemical Name or Material 3-(4-Chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.