missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminobenzoic acid, 94%

Product Code. 11472297
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472297 25 g 25g
11482297 100 g 100g
11492297 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472297 Supplier Thermo Scientific Alfa Aesar Supplier No. A14213.14
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 619-05-6 | C7H8N2O2 | 152.153 g/mol

3,4-Diaminobenzoic acid undergoes cyclocondensations to form, for example, quinoxalines1 and benzimidazoles. It acts as a redox label for electrochemical detection of single base mismatches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Diaminobenzoic acid undergoes cyclocondensations to form, for example, quinoxalines1 and benzimidazoles. It acts as a redox label for electrochemical detection of single base mismatches.

Solubility
Soluble in water (2.2 mg/ml at 20°C), DMF, and methanol (10 mg/ml).

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-05-6
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007726
InChI Key HEMGYNNCNNODNX-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-diamino, 4-carboxyphenyldiamine, 3,4-diamino benzoic acid, 4-carboxy-o-phenylenediamine, 3,4-diaminobenzoicacid, acmc-209mxf, 3,4-diaminobenzoesaeure, 3,4 diaminobenzoic acid, 3,4-diaminobenzenoic acid, 3,4-diamino-benzoic acid
PubChem CID 69263
IUPAC Name 3,4-diaminobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)N

Specifications

Melting Point ∼210°C (decomposition)
Quantity 25 g
Beilstein 775892
Solubility Information Soluble in water (2.2mg/ml at 20°C),DMF,and methanol (10mg/ml).
Formula Weight 152.15
Percent Purity 94%
Chemical Name or Material 3,4-Diaminobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.