missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminobenzophenone, 99%

Product Code. 10792801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10792801 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10792801 Supplier Thermo Scientific Acros Supplier No. 184800250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39070-63-8 | C13H12N2O | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Spécification

CAS 39070-63-8
CAS Min % 98.5
CAS Max % 100.0
Melting Point 116.0°C to 119.0°C
Color Ochre-Yellow to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Molecular Formula C13H12N2O
Linear Formula C6H5COC6H3(NH2)2
MDL Number MFCD00007727
Quantity 25 g
Beilstein 14, II, 225
Synonym 3,4-diaminobenzophenone, 3,4-diaminophenyl phenyl methanone, 3,4-diaminophenyl phenylmethanone, 4-benzoylbenzene-1,2-diamine, 2-amino-4-benzoylaniline, 4-benzoyl-o-phenylenediamine, 3,4-diaminophenyl phenyl ketone, 3,4-diaminophenyl-phenylmethanone, methanone, 3,4-diaminophenyl phenyl, pubchem3382
Solubility Information Solubility in water: 410 mg/l (20°C). Other solubilities: 11g/100mL methanol - 4.7g/100mL ethanol, 1.8g/100mL 2-propanol - >50g/100mL n,n-dmf, 4.7g/100mL methylene chloride - 30g/100mL acetone
InChI Key RXCOGDYOZQGGMK-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)N)N
IUPAC Name (3,4-diaminophenyl)-phenylmethanone
Molecular Weight (g/mol) 212.25
PubChem CID 135520
Formula Weight 212.25
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3, 4-Diaminobenzophenone, 99%
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 3,4-Diaminobenzophenone
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.