missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminotoluene, 97%

Product Code. 11495807
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495807 100 g 100g
11405817 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495807 Supplier Thermo Scientific Alfa Aesar Supplier No. B24378.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 496-72-0 | C7H10N2 | 122.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-72-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00007728
InChI Key DGRGLKZMKWPMOH-UHFFFAOYSA-N
Synonym 3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene
PubChem CID 10332
IUPAC Name 4-methylbenzene-1,2-diamine
SMILES CC1=CC(=C(C=C1)N)N

Specifications

Melting Point 86°C to 90°C
Boiling Point 265°C
Quantity 100 g
Beilstein 507965
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 3,4-Diaminotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.