accessibility menu, dialog, popup

UserName

CAS RN 496-72-0

IUPAC Name: 4-methylbenzene-1,2-diamine
Synonyms: 3,4-diaminotoluene
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: DGRGLKZMKWPMOH-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(N)=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)