missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminotoluene, 97%

Product Code. 10316370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10316370 100 g Glass bottle 100g
10696354 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10316370 Supplier Thermo Scientific Supplier No. 112431000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 496-72-0 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-72-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007728
InChI Key DGRGLKZMKWPMOH-UHFFFAOYSA-N
Synonym 3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene
PubChem CID 10332
IUPAC Name 4-methylbenzene-1,2-diamine
SMILES CC1=CC(=C(C=C1)N)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 83.0°C to 90.0°C
Color Brown to Brown-Gray
Boiling Point 265.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NH2)2
Quantity 100 g
Beilstein 13, 148
Solubility Information Solubility in water: 16g/L (20°C).
Formula Weight 122.17
Percent Purity 97%
Physical Form Crystalline Chunks or Flakes
Chemical Name or Material 3, 4-Diaminotoluene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4-Diaminotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.