missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminotoluene, TRC

Product Code. 30368711
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368711 10 g 10g
30358790 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30368711 Supplier Toronto Research Chemicals Supplier No. TRCD41681010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-72-0
Molecular Formula C7 H10 N2
Molecular Weight (g/mol) 122.17
Synonym Toluene-3,4-diamine (7CI,8CI), 4-Methyl-1,2-benzenediamine, 1,2-Diamino-4-methylbenzene, 2-Amino-4-methylaniline, 2-Amino-4-methylphenylamine, 2-Amino-5-methylaniline, 3,4-Diamino-1-methylbenzene, 3,4-Diaminotoluene, 3,4-Tolylenediamine, 4-Methyl-1,2-diaminobenzene, 4-Methyl-1,2-phenylenediamine, 4-Methyl-2-aminoaniline, 4-Methyl-o-phenylenediamine, 5-Methylbenzene-1,2-diamine, NSC 1495
IUPAC Name 4-methylbenzene-1,2-diamine
SMILES Cc1ccc(N)c(N)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H10N2/c1-5-2-3-6(8)7(9)4-5/h2-4H,8-9H2,1H3
Formula Weight 122.0844
Chemical Name or Material 3,4-Diaminotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.