missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloro Trazodone Monohydrochloride, TRC

Product Code. 30005393
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005393 250 mg 250mg
30007494 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005393 Supplier Toronto Research Chemicals Supplier No. TRCD466655250MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1263278-79-0
Molecular Formula C19H21Cl2N5O . HCl
Molecular Weight (g/mol) 442.8
Synonym 2-[3-[4-(3,4-Dichlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one Monohydrochloride;
IUPAC Name 2-[3-[4-(3,4-dichlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
SMILES Cl.Clc1ccc(cc1Cl)N2CCN(CCCN3N=C4C=CC=CN4C3=O)CC2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H21Cl2N5O.ClH/c20-16-6-5-15(14-17(16)21)24-12-10-23(11-13-24)7-3-9-26-19(27)25-8-2-1-4-18(25)22-26;/h1-2,4-6,8,14H,3,7,9-13H2;1H
Formula Weight 441.089
Chemical Name or Material 3,4-Dichloro Trazodone Monohydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.