missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloroaniline 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30319577
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319577 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319577 Supplier Dr. Ehrenstorfer Supplier No. DREXA12323400AL
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dichloroaniline
CAS 95-76-1
Analyte or Component Names 3,4-Dichloroaniline
Shipping Condition Room Temperature
Molecular Formula C6 H5 Cl2 N
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Aniline, 3,4-dichloro- (7CI,8CI), 3,4-Dichlorobenzenamine, 3,4-DCA, 3,4-Dichloraniline, 3,4-Dichloroaniline, 3,4-Dichlorophenylamine, 4,5-Dichloroaniline, 4-Amino-1,2-dichlorobenzene, DCA, DCA (amine), LY 004892, NSC 247, m,p-Dichloroaniline
InChI Formula InChI=1S/C6H5Cl2N/c7-5-2-1-4(9)3-6(5)8/h1-3H,9H2
Solution Type Single Solution
SMILES Nc1ccc(Cl)c(Cl)c1
IUPAC Name 3,4-dichloroaniline
Solvent or Matrix Acetonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 162.02
Quantity 1 mL
Formula Weight 160.9799 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.