missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloroanisole, 99+%, Thermo Scientific™

Product Code. 10432631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10432631 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10432631 Supplier Thermo Scientific Acros Supplier No. 264190250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 36404-30-5
CAS Min % 99.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C7H6Cl2O
Linear Formula Cl2C6H3OCH3
Refractive Index 1.5560 to 1.558
MDL Number MFCD00070785
Quantity 25 g
Synonym 3,4-dichloroanisole, benzene, 1,2-dichloro-4-methoxy, 3,4-dichloromethoxybenzene, 1,2-dichloro-4-methyxybenzene, 3,4-dichloroanizole, pubchem3683, acmc-1ajco, 4,5-dichlorophenyl methyl ether, 3,4-dichlorophenol, methyl ether
InChI Key VISJRVXHPNMYRH-UHFFFAOYSA-N
SMILES COC1=CC(=C(C=C1)Cl)Cl
IUPAC Name 1,2-dichloro-4-methoxybenzene
Molecular Weight (g/mol) 177.03
PubChem CID 37438
Formula Weight 177.03
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3, 4-Dichloroanisole, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichloroanisole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.