missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzene-1-carbohydrazide, 97%, Thermo Scientific™

Product Code. 10520363
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10520363 1 g Amber Glass Bottle 1g
10160562 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10520363 Supplier Thermo Scientific Maybridge Supplier No. TL00776DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 28036-91-1
Molecular Formula C7H6Cl2N2O
Molecular Weight (g/mol) 205.038
MDL Number MFCD00017057
InChI Key VEDIADQJSGNNQE-UHFFFAOYSA-N
PubChem CID 311996
IUPAC Name 3,4-dichlorobenzohydrazide
SMILES C1=CC(=C(C=C1C(=O)NN)Cl)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3C(O)NHNH2
Quantity 1 g
Synonym 3,4-dichlorobenzene-1-carbohydrazide, 3,4-dichlorobenzenecarbohydrazide, 3,4-dichlorobenzoic acid hydrazide, 3,4-dichlorobenzhydrazide, dichlorobenzenecarbohydrazide, 3,4-dichlor-benzoyl-hydrazin, 3,4-dichlorobenzoic hydrazide, 1,2-dichlorobenzene-4-carbohydrazide, benzoic acid,3,4-dichloro-, hydrazide, benzoic acid, 3,4-dichloro-, hydrazide
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material 3, 4-Dichlorobenzene-1-carbohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.