missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzoic acid, 98+%, ACROS Organics™

Product Code. 10453083 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10453083 25 g Plastic bottle 25g
10168612 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10453083 Supplier Thermo Scientific Acros Supplier No. 374210250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51-44-5
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002492
InChI Key VPHHJAOJUJHJKD-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609
PubChem CID 5817
ChEBI CHEBI:49401
IUPAC Name 3,4-dichlorobenzoic acid
SMILES OC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 205.0°C to 209.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula Cl2C6H3CO2H
Quantity 25 g
Formula Weight 191.01
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 3, 4-Dichlorobenzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.