missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzoic acid, 99%

Product Code. p-7022899
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11402588

Brand: Thermo Scientific Alfa Aesar A15873.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51-44-5 | C7H4Cl2O2 | 191.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-44-5
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002492
InChI Key VPHHJAOJUJHJKD-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609
PubChem CID 5817
ChEBI CHEBI:49401
IUPAC Name 3,4-dichlorobenzoic acid
SMILES OC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 205°C to 208°C
Quantity 25 g
Beilstein 2044777
Formula Weight 191.01
Percent Purity 99%
Chemical Name or Material 3,4-Dichlorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.