missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl chloride, 98+%

Product Code. 10512081 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10512081

Brand: Thermo Scientific Acros 279051000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 102-47-6 | C7H5Cl3 | 195.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-47-6
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000906
InChI Key YZIFVWOCPGPNHB-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl chloride, 1,2-dichloro-4-chloromethyl benzene, alpha,3,4-trichlorotoluene, benzene, 1,2-dichloro-4-chloromethyl, unii-451pb8smie, 1,2-dichloro-4-chloromethyl-benzene, 3,4-dichlorobenzylchloride, 451pb8smie, 3,4-dichlorophenylmethyl chloride, toluene, .alpha.,3,4-trichloro
PubChem CID 7609
IUPAC Name 1,2-dichloro-4-(chloromethyl)benzene
SMILES C1=CC(=C(C=C1CCl)Cl)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -3°C
Color Colorless to Yellow
Density 1.4100g/mL
Boiling Point 122°C to 124°C (14.0 mmHg)
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3CH2Cl
Refractive Index 1.5775
Quantity 100 g
Beilstein 05, 300
Specific Gravity 1.41
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 195.47
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 3, 4-Dichlorobenzyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4-Dichlorobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.