Learn More
3,4-Dichlorophenol, 97%
Description
It is also a photo-degradation product of the common antibacterial and antifungal agent triclosan along with the dioxin 2,8-dichlorodibenzo-p-dioxin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is also a photo-degradation product of the common antibacterial and antifungal agent triclosan along with the dioxin 2,8-dichlorodibenzo-p-dioxin.
Solubility
Slightly soluble ion water 9.2 g/L at 20degC. Soluble in aqueous alkali, oxygenated and chlorinated solvents. Soluble in ethanol, benzene, chloroform and ethyl ether.
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents, acid, acid fumes, acid chlorides, acid anhydrides, caustics. Keep container tightly closed. Store in an area without drain or sewer access. Separated from strong oxidants and food and feedstuffs.
Specifications
Specifications
| Melting Point | 64°C to 68°C |
| Boiling Point | 252°C to 255°C |
| Odor | Phenol-like |
| Quantity | 100 g |
| UN Number | UN2020 |
| Beilstein | 1907693 |
| Synonym | 4,5-dichlorophenol, phenol, 3,4-dichloro, 3,3-dichloro-hydroxybenzene, unii-070ftm6dvf, ccris 5904, 070ftm6dvf, 3,4-dcp, 3,4-dichlorphenol, 3,4dichlorophenol, 3,4 dichlorophenol |
| Solubility Information | Slightly soluble ion water 9.2g/L at 20degC. Soluble in aqueous alkali,oxygenated and chlorinated solvents. Soluble in ethanol,benzene,chloroform and ethyl ether. |
| Formula Weight | 163 |
| Percent Purity | 99% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.