missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzaldehyde, 96%

Product Code. 10645701 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10645701 25 g Glass bottle 25g
10114490 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10645701 Supplier Thermo Scientific Acros Supplier No. 271300250
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 34036-07-2 | C7H4F2O | 142.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34036-07-2
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.1
MDL Number MFCD00010328
InChI Key JPHKMYXKNKLNDF-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-difluoro, pubchem1413, 3,4 difluorobenzaldehyde, 3,4-diflourobenzaldehyde, 3.4-difluorobenzaldehyde, 3,4-difluoro-benzaldehye, 3,4-difluro-benzaldehyde, acmc-1ajc9, 3, 4-difluorobenzaldehyde, 3,4-difluoro benzaldehyde
PubChem CID 588088
IUPAC Name 3,4-difluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)F)F

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2900g/mL
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3CHO
Refractive Index 1.4990 to 1.5010
Quantity 25 g
Specific Gravity 1.29
Formula Weight 142.1
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3, 4-Difluorobenzaldehyde, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Difluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.