missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzaldehyde, 98%, Thermo Scientific™

Product Code. 11497906
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497906 25 g 25g
11487906 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497906 Supplier Thermo Scientific Alfa Aesar Supplier No. A14028.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34036-07-2
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.105
MDL Number MFCD00010328
InChI Key JPHKMYXKNKLNDF-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-difluoro, pubchem1413, 3,4 difluorobenzaldehyde, 3,4-diflourobenzaldehyde, 3.4-difluorobenzaldehyde, 3,4-difluoro-benzaldehye, 3,4-difluro-benzaldehyde, acmc-1ajc9, 3, 4-difluorobenzaldehyde, 3,4-difluoro benzaldehyde
PubChem CID 588088
IUPAC Name 3,4-difluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)F)F

Specifications

Density 1.288
Boiling Point 180°C
Flash Point 69°C (156°F)
Refractive Index 1.5
Quantity 25 g
Beilstein 2241231
Sensitivity Air Sensitive
Formula Weight 142.11
Percent Purity 98%
Chemical Name or Material 3,4-Difluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.