missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzoic acid, 98%

Product Code. p-7022900
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11452588

Brand: Thermo Scientific Alfa Aesar A15876.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 455-86-7 | C7H4F2O2 | 158.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-86-7
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00011672
InChI Key FPENCTDAQQQKNY-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-difluoro, 3,4-fifluorobenzoic acid, 3,4-difluoro-benzoic acid, 3,4-dichloropnenol, pubchem1337, acmc-209k3h, 3,4-difluoro benzoic acid, ksc237g8t, rarechem al bo 0269, timtec-bb sbb006721
PubChem CID 99166
IUPAC Name 3,4-difluorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)F)F

Specifications

Melting Point 121°C to 124°C
Odor Odorless
Quantity 1 g
Beilstein 2085848
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material 3,4-Difluorobenzoic acid
compliance-icons
Product Identifier
  • 3,4-Difluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 3,4-Difluorobenzoicacid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.