missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzotrifluoride, 97%, Thermo Scientific Chemicals

Product Code. 11421107
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421107 5 g 5g
11411107 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11421107 Supplier Thermo Scientific Alfa Aesar Supplier No. B24096.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32137-19-2
Molecular Formula C7H3F5
Molecular Weight (g/mol) 182.09
MDL Number MFCD00077510
InChI Key MVCGQTYWLZSKSB-UHFFFAOYSA-N
Synonym 3,4-difluorobenzotrifluoride, 1,2-difluoro-4-trifluoromethyl benzene, alpha,alpha,alpha,3,4-pentafluorotoluene, benzene, 1,2-difluoro-4-trifluoromethyl, pubchem1651, ksc497k4r, 1,2-difluoro-4-trifluoromethyl benzene #
PubChem CID 602797
IUPAC Name 1,2-difluoro-4-(trifluoromethyl)benzene
SMILES FC1=CC=C(C=C1F)C(F)(F)F

Specifications

Boiling Point 103°C to 104°C
Refractive Index 1.3915
Quantity 5 g
UN Number UN1993
Beilstein 1950149
Formula Weight 182.1
Percent Purity 97%
Chemical Name or Material 3,4-Difluorobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.