missing translation for 'onlineSavingsMsg'
Learn More

3,4-difluorobenzyl Chloride, 98%

Product Code. 10699984 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10699984

Brand: Thermo Scientific Acros 384420250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 698-80-6 | C7H5ClF2 | 162.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-80-6
Molecular Formula C7H5ClF2
Molecular Weight (g/mol) 162.56
MDL Number MFCD00236218
InChI Key VTGRVYJPIVZZGS-UHFFFAOYSA-N
Synonym 3,4-difluorobenzyl chloride, 4-chloromethyl-1,2-difluorobenzene, 3,4-difluorobenzylchloride, benzene, 4-chloromethyl-1,2-difluoro, 1-chloromethyl-3,4-difluorobenzene, alpha-chloro-3,4-difluorotoluene, 3, 4-difluorobenzyl chloride, pubchem7357, 4-chloromethyl-1,2-difluoro-benzene
PubChem CID 2736954
IUPAC Name 4-(chloromethyl)-1,2-difluorobenzene
SMILES FC1=CC=C(CCl)C=C1F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2C6H3F2
Refractive Index 1.4920 to 1.4940 (20°C, 589nm)
Quantity 25 g
Formula Weight 162.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 4-Difluorobenzyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4-Difluorobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.