missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenol, 97%, Thermo Scientific™

Product Code. 10113364
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
Unit Size:
25g
Product Code. Quantity Packaging unitSize
10113364 25 g Amber Glass Bottle 25g
1 options
This item is not returnable. View return policy

Product Code. 10113364

Brand: Thermo Scientific Maybridge SB01786EB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 2713-33-9
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H4F2O
Linear Formula F2(C6H3)OH
MDL Number MFCD00010315
Quantity 25 g
Synonym phenol, 3,4-difluoro, 3,4-difluoro phenol, 3 4-difluorophenol, 3,4-difluoro-phenol, pubchem2315, acmc-1cmaj, 3,4-difluorophenol, ksc202q5j, bnpwvujopcghik-uhfffaoysa
InChI Key BNPWVUJOPCGHIK-UHFFFAOYSA-N
SMILES OC1=CC=C(F)C(F)=C1
IUPAC Name 3,4-difluorophenol
Molecular Weight (g/mol) 130.09
PubChem CID 75927
Formula Weight 130.09
Percent Purity 97%
Chemical Name or Material 3, 4-Difluorophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.