missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorosalicylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11322049
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322049 1 g 1g
15463367 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322049 Supplier Thermo Scientific Alfa Aesar Supplier No. H26088.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 189283-51-0
Molecular Formula C7H4F2O3
Molecular Weight (g/mol) 174.103
MDL Number MFCD03428564
InChI Key GWOOBUWKTOCYKY-UHFFFAOYSA-N
Synonym 3,4-difluorosalicylic acid, benzoic acid, 3,4-difluoro-2-hydroxy, 3,4-difluoro-2-hydroxy-benzoic acid, acmc-20anf8, 3,4-difluoro-hydroxybenzoic acid, 3,4-difluro-2-hydroxybenzoic acid, 3,4-difluoro-2-hydroxy benzoic acid, 3,4-bis fluoranyl-2-oxidanyl-benzoic acid
PubChem CID 2758286
IUPAC Name 3,4-difluoro-2-hydroxybenzoic acid
SMILES C1=CC(=C(C(=C1C(=O)O)O)F)F

Spezifikation

Melting Point 177°C to 179°C
Quantity 1 g
Formula Weight 174.1
Percent Purity 98%
Chemical Name or Material 3,4-Difluorosalicylic acid

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.