missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde, 95%

Product Code. 15239686 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15239686 1 g 1g
15249686 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15239686 Supplier Thermo Scientific Acros Supplier No. 450440010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 67869-90-3 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67869-90-3
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
InChI Key LCSVYSVGXQQHSI-UHFFFAOYSA-N
PubChem CID 2776388
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepine-7-carbaldehyde
SMILES C1COC2=C(C=C(C=C2)C=O)OC1

Specifications

Boiling Point 120.0°C to 122.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym 2h-1,5-benzodioxepin-7-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxaldehyde, 2h-1,5-benzodioxepin-7-carboxaldehyde,3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepane-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepine-7-carboxaldehyde, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyd
Formula Weight 178.19
Percent Purity 95%
Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.