missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxycinnamic acid, predominantly trans, 98+%

Product Code. 11498496
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498496 10 g 10g
11408506 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498496 Supplier Thermo Scientific Alfa Aesar Supplier No. A15950.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 331-39-5 | C9H8O4 | 180.159 g/mol

3,4-Dihydroxycinnamic acid, predominantly trans stimulates prostaglandin synthesis at high doses. It has antioxidant, anti-inflammatory, anti-tumor and immunomodulatory properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxycinnamic acid, predominantly trans stimulates prostaglandin synthesis at high doses. It has antioxidant, anti-inflammatory, anti-tumor and immunomodulatory properties.

Solubility
Soluble in DMSO (40 mg/mol), ethanol (25 mg/mL warm), DMF (∼7 mg/mL), PBS, PH 7.2 (∼0.65 mg/mol), hot water, ethyl acetate, Chloroform Hexane, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331-39-5
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00004392
InChI Key QAIPRVGONGVQAS-DUXPYHPUSA-N
Synonym caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid
PubChem CID 689043
ChEBI CHEBI:16433
IUPAC Name (E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1C=CC(=O)O)O)O

Specifications

Melting Point ∼202°C (decomposition)
Quantity 10 g
Beilstein 2210883
Merck Index 14,1635
Solubility Information Soluble in DMSO (40mg/mol),ethanol (25mg/mL warm),DMF (∽7mg/mL),PBS,PH 7.2 (∽0.65mg/mol),hot water,ethyl acetate,Chloroform Hexane,and methanol.
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material 3,4-Dihydroxycinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.