missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimercaptotoluene, 95%

Product Code. 10750872 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10750872 1 g Glass bottle 1g
10361401 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10750872 Supplier Thermo Scientific Acros Supplier No. 421180010
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 496-74-2 | C7H8S2 | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-74-2
Molecular Formula C7H8S2
Molecular Weight (g/mol) 156.27
MDL Number MFCD00004844
InChI Key NIAAGQAEVGMHPM-UHFFFAOYSA-N
Synonym toluene-3,4-dithiol, dithiol, 1,2-benzenedithiol, 4-methyl, 3,4-dimercaptotoluene, 3,4-dimercaptotoluol, 1,2-dimercapto-4-methylbenzene, 4-methyl-1,2-benzenedithiol, 3,4-toluenedithiol, o-toluenesulfonylamide, usaf b-59
PubChem CID 10334
IUPAC Name 4-methylbenzene-1,2-dithiol
SMILES CC1=CC(=C(C=C1)S)S

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 27°C
Color White to Yellow
Density 1.1800g/mL
Boiling Point 185°C (84.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 9690
Specific Gravity 1.18
Formula Weight 156.27
Percent Purity 95%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 3, 4-Dimercaptotoluene, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimercaptotoluene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.