missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzoic acid, 99+%

Product Code. 11469452
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469452 250 g 250g
11459452 50 g 50g
11449452 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469452 Supplier Thermo Scientific Alfa Aesar Supplier No. A10167.30
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-07-2 | C9H10O4 | 182.175 g/mol

3,4-Dimethoxybenzoic acid plays an important role in producing antibiotics and various dyes. They are used as intermediates for pharmaceuticals (especially for antipyretic analgesic, antirheumatism) and other organic synthesis. It is used as matrix for ionization of peptides, proteins and carbohydrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxybenzoic acid plays an important role in producing antibiotics and various dyes. They are used as intermediates for pharmaceuticals (especially for antipyretic analgesic, antirheumatism) and other organic synthesis. It is used as matrix for ionization of peptides, proteins and carbohydrates.

Solubility
Soluble in water (0.5 mg/ml at 15°C), chloroform, methanol, and 95% ethanol (50 mg/ml).

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-07-2
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00002500
InChI Key DAUAQNGYDSHRET-UHFFFAOYSA-N
Synonym veratric acid, benzoic acid, 3,4-dimethoxy, dimethylprotocatechuic acid, veratrylic acid, 3,4-dimethylprotocatechuic acid, veratrumenoic acid, 3,4-dimethoxy-benzoic acid, unii-1yy04e7rr4, 3,4-dimethoxybenzoes
PubChem CID 7121
ChEBI CHEBI:296881
IUPAC Name 3,4-dimethoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)OC

Specifications

Melting Point 179°C to 183°C
Odor Odorless
Quantity 250 g
Beilstein 518285
Merck Index 14,9953
Solubility Information Soluble in water (0.5mg/ml at 15°C),chloroform,methanol,and 95% ethanol (50mg/ml).
Formula Weight 182.18
Percent Purity ≥99%
Chemical Name or Material 3,4-Dimethoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.