missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzoic acid, 99+%

Product Code. 10752254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10752254 25 g Glass bottle 25g
10080760 100 g Glass bottle 100g
10529243 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10752254 Supplier Thermo Scientific Acros Supplier No. 115450250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-07-2 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-07-2
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002500
InChI Key DAUAQNGYDSHRET-UHFFFAOYSA-N
Synonym veratric acid, benzoic acid, 3,4-dimethoxy, dimethylprotocatechuic acid, veratrylic acid, 3,4-dimethylprotocatechuic acid, veratrumenoic acid, 3,4-dimethoxy-benzoic acid, unii-1yy04e7rr4, 3,4-dimethoxybenzoes
PubChem CID 7121
ChEBI CHEBI:296881
IUPAC Name 3,4-dimethoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)OC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 179°C to 182°C
Color Beige to White or Red
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CO2H
Quantity 25 g
Beilstein 10, I, 188
Merck Index 15, 10147
Solubility Information Solubility in water: 0.5g/L (14°C). Other solubilities: readily soluble in alcohol,soluble in ether
Formula Weight 182.18
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dimethoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.