missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxycinnamic Acid, TRC

Product Code. 30383499
Change view
Click to view available options
Quantity:
1 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383499 1 g
30380956 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383499 Supplier Toronto Research Chemicals Supplier No. TRCD4736951G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2316-26-9
Molecular Formula C11 H12 O4
Molecular Weight (g/mol) 208.21
Synonym 3,4-Dimethoxycinnamic acid, 3,4-Dimethoxyphenyl-2-propenoic acid, (E/Z)-Caffeic acid dimethyl ether
IUPAC Name 3-(3,4-dimethoxyphenyl)prop-2-enoic acid
SMILES COc1ccc(C=CC(=O)O)cc1OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H12O4/c1-14-9-5-3-8(4-6-11(12)13)7-10(9)15-2/h3-7H,1-2H3,(H,12,13)
Formula Weight 208.0736
Chemical Name or Material Dimethylcaffeic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.