missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylaniline, 98%

Product Code. 11369529
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369529 100 g 100g
11458756 250 g 250g
11448756 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369529 Supplier Thermo Scientific Alfa Aesar Supplier No. A16928.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 95-64-7 | C8H11N | 121.18 g/mol

3,4-Dimethylaniline is used to study electron donor-acceptor interaction with DCNQ. It is also used as a fine chemical &organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylaniline is used to study electron donor-acceptor interaction with DCNQ. It is also used as a fine chemical &organic intermediate.

Solubility
Soluble in water and in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates, halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-64-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007810
InChI Key DOLQYFPDPKPQSS-UHFFFAOYSA-N
Synonym 3,4-xylidine, 4-amino-o-xylene, 3,4-xylylamine, 3,4-dimethylphenylamine, benzenamine, 3,4-dimethyl, 3,4-dimethylbenzenamine, 3,4-dimethylaminobenzene, 1-amino-3,4-dimethylbenzene, 4-amino-1,2-dimethylbenzene, aniline, 3,4-dimethyl
PubChem CID 7248
ChEBI CHEBI:39901
IUPAC Name 3,4-dimethylaniline
SMILES CC1=CC=C(N)C=C1C

Specifications

Melting Point 48°C to 52°C
Density 1.076
Quantity 100 g
Odor Characteristic
UN Number UN3452
Beilstein 507414
Merck Index 14,10084
Boiling Point 226°C to 228°C
Solubility Information Soluble in water and in methanol.
Formula Weight 121.18
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.