missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylthiophenol, 98%

Product Code. 11420757
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420757 1 g 1g
11430757 5 g 5g
11440757 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420757 Supplier Thermo Scientific Alfa Aesar Supplier No. B23168.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 18800-53-8 | C8H10S | 138.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18800-53-8
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010023
InChI Key IDKCKPBAFOIONK-UHFFFAOYSA-N
Synonym 3,4-dimethylthiophenol, benzenethiol, 3,4-dimethyl, 3,4-dimethyl thiophenol, o-xylene-4-thiol, 4-mercapto-o-xylene, 3,4-dimethylbenzene-1-thiol, pubchem6812, 3,4-dimethyl-thiophenol, acmc-1bu1r, 3,4-dimethyl-benzenethiol
PubChem CID 87799
IUPAC Name 3,4-dimethylbenzenethiol
SMILES CC1=C(C=C(C=C1)S)C

Specifications

Density 1.027
Boiling Point 218°C
Flash Point 90°C (194°F)
Odor Unpleasant
Refractive Index 1.5736
Quantity 1 g
UN Number UN2810
Beilstein 2040353
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylthiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.