missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylthiophenol, 98%, Thermo Scientific™

Product Code. 11965741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11965741 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11965741 Supplier Thermo Scientific Acros Supplier No. 307350010
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 18800-53-8
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0270g/mL
Boiling Point 218°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H10S
Linear Formula CH3C6H3SHCH3
Refractive Index 1.5730 to 1.5750 (20°C, 589nm)
MDL Number MFCD00010023
Quantity 1 g
Specific Gravity 1.027
Synonym 3,4-dimethylthiophenol, benzenethiol, 3,4-dimethyl, 3,4-dimethyl thiophenol, o-xylene-4-thiol, 4-mercapto-o-xylene, 3,4-dimethylbenzene-1-thiol, pubchem6812, 3,4-dimethyl-thiophenol, acmc-1bu1r, 3,4-dimethyl-benzenethiol
InChI Key IDKCKPBAFOIONK-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)S)C
IUPAC Name 3,4-dimethylbenzenethiol
Molecular Weight (g/mol) 138.23
PubChem CID 87799
Formula Weight 138.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 4-Dimethylthiophenol, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.