missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15497985
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497985 5 g 5g
15487985 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497985 Supplier Thermo Scientific Alfa Aesar Supplier No. H32090.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1736-21-6 ∣ C11H8FNO3 ∣ 221.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1736-21-6
Molecular Formula C11H8FNO3
Molecular Weight (g/mol) 221.19
MDL Number MFCD03407356
InChI Key PDEGBONVUJDOFN-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-4-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, 3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-4-fluorophenyl-5-methyl, 3-4-fluorophenyl-5-methyl-4-isoxazolecarboxylicacid, 3-4-fluorophenyl-5-methylisoxazol-4-carboxylic acid, buttpark 9912-50, 4-isoxazolecarboxylicacid, 3-4-fluorophenyl-5-methyl, 4-isoxazolecarboxylicacid,3-4-fluorophenyl-5-methyl, 3-4-fluoro-phenyl-5-methyl-isoxazole-4-carboxylic acid
PubChem CID 2780709
IUPAC Name 3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=CC=C(F)C=C1

Specifications

Melting Point 199°C to 201°C
Quantity 5 g
Sensitivity Light and moisture sensitive
Formula Weight 221.19
Percent Purity 97%
Chemical Name or Material 3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.