missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxy-3-methoxyphenyl)propionic acid, 97%

Product Code. 11489962
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489962 5 g 5g
11479962 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489962 Supplier Thermo Scientific Alfa Aesar Supplier No. A12069.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1135-23-5 | C10H12O4 | 196.202 g/mol

It is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-23-5
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00016558
InChI Key BOLQJTPHPSDZHR-UHFFFAOYSA-N
Synonym 3-4-hydroxy-3-methoxyphenyl propanoic acid, 3-4-hydroxy-3-methoxyphenyl propionic acid, dihydroferulic acid, hydroferulic acid, dihydroconiferylic acid, unii-o01rnc700m, benzenepropanoic acid, 4-hydroxy-3-methoxy, 3-4-hydroxymethyl propionic acid, .beta.-4-hydroxy-3-methoxyphenyl propionic acid, 3-4-hydroxy-3-methoxy-phenyl propanoic acid
PubChem CID 14340
ChEBI CHEBI:86612
IUPAC Name 3-(4-hydroxy-3-methoxyphenyl)propanoic acid
SMILES COC1=C(C=CC(=C1)CCC(=O)O)O

Specifications

Melting Point 87°C to 92°C
Quantity 5 g
Beilstein 2110370
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 196.2
Percent Purity 97%
Chemical Name or Material 3- (4-Hydroxy-3-methoxyphenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.