missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Hydroxyphenyl)propionic acid, 99%
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Beskrivning
3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifikationer
Specifikationer
| Melting Point | 126°C to 131°C |
| Quantity | 10 g |
| Beilstein | 2209841 |
| Solubility Information | Soluble in water. |
| Formula Weight | 166.18 |
| Percent Purity | 99% |
| Chemical Name or Material | 3-(4-Hydroxyphenyl)propionic acid |
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering