accessibility menu, dialog, popup

UserName

CAS RN 501-97-3

IUPAC Name: 3-(4-hydroxyphenyl)propanoic acid
Synonyms: hydroxyphenyl propionic acid 4-(4-hydroxyphenylpropanoate hydro-p-coumaric acid phloretic acid hydro-p-coumarate dihydro-p-coumarate phloretinate P-hydroxy-B-propionic acid desaminotyrosine uhloretic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: NMHMNPHRMNGLLB-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)