missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)-2-propynoic Acid, TRC

Product Code. 30477588
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477588 100 mg 100mg
30476466 50 mg 50mg
30480974 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477588 Supplier Toronto Research Chemicals Supplier No. TRCM228255100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2227-57-8
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
Synonym 3-(4-Methoxyphenyl)propiolic acid
IUPAC Name 3-(4-methoxyphenyl)prop-2-ynoic acid
SMILES COc1ccc(cc1)C#CC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H8O3/c1-13-9-5-2-8(3-6-9)4-7-10(11)12/h2-3,5-6H,1H3,(H,11,12)
Formula Weight 176.0473
Chemical Name or Material 3-(4-Methoxyphenyl)-2-propynoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.